missing translation for 'onlineSavingsMsg'
Learn More

N-Fluoropyridinium trifluoromethanesulfonate, 95%

Catalog No. aal1432403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1432403 1 g
AAL14324MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1432403 Supplier Thermo Scientific Chemicals Supplier No. L1432403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107263-95-6 | C6H5F4NO3S | 247.164 g/mol

N-Fluoropyridinium trifluoromethanesulfonate is used as a reagent for electrophilic fluorination and for a review of electrophilic N-F fluorinating agents. It is also used as a reactant for Pd-catalyzed arylation reactions and as a fluorination agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fluoropyridinium trifluoromethanesulfonate is used as a reagent for electrophilic fluorination and for a review of electrophilic N-F fluorinating agents. It is also used as a reactant for Pd-catalyzed arylation reactions and as a fluorination agent.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with oxidizing agents. Store under dry inert gas.

Chemical Identifiers

CAS 107263-95-6
Molecular Formula C6H5F4NO3S
Molecular Weight (g/mol) 247.164
MDL Number MFCD00013458
InChI Key JFZMMCYRTJBQQI-UHFFFAOYSA-M
Synonym 1-fluoropyridinium triflate, n-fluoropyridinium triflate, n-fluoropyridinium trifluoromethanesulfonate, 1-fluoropyridinium trifluoromethanesulfonate, 1-fluoropyridin-1-ium; trifluoromethanesulfonate, acmc-209shu, ksc182s3j, 1-fluoropyridin-1-ium triflate, 1-fluoropyridiniumtriflate
PubChem CID 2724576
IUPAC Name 1-fluoropyridin-1-ium;trifluoromethanesulfonate
SMILES C1=CC=[N+](C=C1)F.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 180°C to 185°C
Quantity 1 g
UN Number UN3261
Beilstein 4222821
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 247.17
Percent Purity 95%
Chemical Name or Material N-Fluoropyridinium trifluoromethanesulfonate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only