CAS RN 107263-95-6
IUPAC Name:
1-fluoropyridin-1-ium trifluoromethanesulfonate
Synonyms:
N-fluoropyridinium triflate
Molecular Weight (g/mol):
247.16
Molecular Formula:
C6H5F4NO3S
InChi Key:
JFZMMCYRTJBQQI-UHFFFAOYSA-M
SMILES:
F[N+]1=CC=CC=C1.[O-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
N-Fluoropyridinium trifluoromethanesulfonate, 95%
N-Fluoropyridinium trifluoromethanesulfonate, 95%
CAS: 107263-95-6 Molecular Formula: C6H5F4NO3S Molecular Weight (g/mol): 247.164 MDL Number: MFCD00013458 InChI Key: JFZMMCYRTJBQQI-UHFFFAOYSA-M Synonym: 1-fluoropyridinium triflate,n-fluoropyridinium triflate,n-fluoropyridinium trifluoromethanesulfonate,1-fluoropyridinium trifluoromethanesulfonate,1-fluoropyridin-1-ium; trifluoromethanesulfonate,acmc-209shu,ksc182s3j,1-fluoropyridin-1-ium triflate,1-fluoropyridiniumtriflate PubChem CID: 2724576 IUPAC Name: 1-fluoropyridin-1-ium;trifluoromethanesulfonate SMILES: C1=CC=[N+](C=C1)F.C(F)(F)(F)S(=O)(=O)[O-]