missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Ethyltoluenesulfonamide (o- and p- mixture) 98.0+%, TCI America™
$46.44 - $139.35
Chemical Identifiers
CAS | 76902-32-4 |
---|---|
Molecular Formula | C18H26N2O4S2 |
Molecular Weight (g/mol) | 398.54 |
MDL Number | MFCD00048511 |
InChI Key | IDLSDSKPHLCUTN-UHFFFAOYSA-N |
Chemical Identifiers
76902-32-4 | |
398.54 | |
IDLSDSKPHLCUTN-UHFFFAOYSA-N |
C18H26N2O4S2 | |
MFCD00048511 |
Specifications
76902-32-4 | |
C18H26N2O4S2 | |
25 g | |
398.54 | |
≥98.0% (N) | |
N-Ethyltoluenesulfonamide (o- and p- mixture) |
Yellow | |
MFCD00048511 | |
IDLSDSKPHLCUTN-UHFFFAOYSA-N | |
199.27 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-1929
TSCA : No