accessibility menu, dialog, popup

UserName

CAS RN 76902-32-4

CH3NHSOOCH3CH3NHSOOH3C

CAS RN 76902-32-4

IUPAC Name: N-ethyl-2-methylbenzene-1-sulfonamide; N-ethyl-4-methylbenzene-1-sulfonamide
Synonyms: N-ethyl-2-methylbenzenesulfonamide; N-ethyl-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 398.54
Molecular Formula: C18H26N2O4S2
InChi Key: IDLSDSKPHLCUTN-UHFFFAOYSA-N
SMILES: CCNS(=O)(=O)C1=CC=C(C)C=C1.CCNS(=O)(=O)C1=CC=CC=C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results