missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylpropylamine, 97+%

Catalog No. p-7049578
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1484606 5 g
AA1484614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1484606 Supplier Thermo Scientific Chemicals Supplier No. 01484606
Only null left

CAS: 20193-20-8 | C5H13N | 87.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20193-20-8
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00015198
InChI Key XCVNDBIXFPGMIW-UHFFFAOYSA-N
Synonym n-ethylpropylamine, 1-propanamine, n-ethyl, ethyl propyl amine, propylamine, n-ethyl, ethylpropylamine, n-ethyl-n-propylamine, ethyl-propyl-amine, n-propylethylamine, n-ethylpropanamine, ethyl-n-propylamine
PubChem CID 88398
IUPAC Name N-ethylpropan-1-amine
SMILES CCCNCC

Specifications

Density 0.748 g/mL
Boiling Point 80°C to 85°C
Flash Point 4.4°C (40°F)
Quantity 5 g
UN Number UN2733
Formula Weight 87.16
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material N-Ethylpropylamine
Hazard Category H225-H302+H332-H311-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 243-573-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only