missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmethylamine 98.0+%, TCI America™

Supplier:  TCI America E072925G

 View more versions of this product

Catalog No. E072925G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Ethylmethylamine
Yellow
C3H9N
25 g
n-ethylmethylamine, methylethylamine, ethanamine, n-methyl, n-methylethylamine, ethyl methyl amine, ethylmethylamine, nmea, n-methyl-n-ethylamine, ethylamine, n-methyl, ethyl-methyl-amine
CCNC
59.11
59.11
Liquid
624-78-2
35°C
MFCD00009030
2733
LIWAQLJGPBVORC-UHFFFAOYSA-N
ethyl(methyl)amine
12219
≥98.0% (GC)

Chemical Identifiers

624-78-2
59.11
LIWAQLJGPBVORC-UHFFFAOYSA-N
12219
CCNC
C3H9N
MFCD00009030
n-ethylmethylamine, methylethylamine, ethanamine, n-methyl, n-methylethylamine, ethyl methyl amine, ethylmethylamine, nmea, n-methyl-n-ethylamine, ethylamine, n-methyl, ethyl-methyl-amine
ethyl(methyl)amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4112

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions