missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmethylamine 98.0+%, TCI America™

Catalog No. p-7129118
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07295G 5 g
E072925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07295G Supplier TCI America Supplier No. E07295G
Only null left

Chemical Identifiers

CAS 624-78-2
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00009030
InChI Key LIWAQLJGPBVORC-UHFFFAOYSA-N
Synonym n-ethylmethylamine, methylethylamine, ethanamine, n-methyl, n-methylethylamine, ethyl methyl amine, ethylmethylamine, nmea, n-methyl-n-ethylamine, ethylamine, n-methyl, ethyl-methyl-amine
PubChem CID 12219
IUPAC Name ethyl(methyl)amine
SMILES CCNC

Specifications

Color Yellow
Boiling Point 35°C
Quantity 5 g
UN Number 2733
Formula Weight 59.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Ethylmethylamine
EINECSNumber (2)-4112
TSCA Yes
Recommended Storage Refrigerator