missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylglycine 98.0+%, TCI America™

Supplier:  TCI America E05175G

 View more versions of this product

Catalog No. E05175G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Ethylglycine
190°C
C4H9NO2
5 g
YPIGGYHFMKJNKV-UHFFFAOYSA-N
2-(ethylamino)acetic acid
316542
103.12
Crystalline Powder
627-01-0
White-Yellow
MFCD00037794
n-ethylglycine, 2-ethylamino acetic acid, n-ethyl glycine, etgly, glycine, n-ethyl, monoethylglycine, pubchem23122, acmc-1au6b, ypiggyhfmkjnkv-uhfffaoysa
CCNCC(=O)O
103.121
CHEBI:15620
≥98.0% (T)

Chemical Identifiers

627-01-0
103.121
YPIGGYHFMKJNKV-UHFFFAOYSA-N
316542
2-(ethylamino)acetic acid
C4H9NO2
MFCD00037794
n-ethylglycine, 2-ethylamino acetic acid, n-ethyl glycine, etgly, glycine, n-ethyl, monoethylglycine, pubchem23122, acmc-1au6b, ypiggyhfmkjnkv-uhfffaoysa
CHEBI:15620
CCNCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions