missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylethylenediamine 99.0+%, TCI America™

Supplier:  TCI America E0294100ML

 View more versions of this product

Catalog No. E0294100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Ethylethylenediamine
Yellow
C4H12N2
100 mL
n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine
CCNCCN
88.154
88.15
Liquid
110-72-5
130°C
MFCD00008166
2734
SCZVXVGZMZRGRU-UHFFFAOYSA-N
N'-ethylethane-1,2-diamine
66071
≥99.0% (GC)

Chemical Identifiers

110-72-5
88.154
SCZVXVGZMZRGRU-UHFFFAOYSA-N
66071
CCNCCN
C4H12N2
MFCD00008166
n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine
N'-ethylethane-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3275

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions