missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-p-toluidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 450781000

Quantity 100g

 View more versions of this product

Catalog No. AC450781000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

622-57-1
135.21
n-ethyl-p-toluidine, benzenamine, n-ethyl-4-methyl, n-ethyl-4-toluidine, n-ethyl-p-methylaniline, p-methyl-n-ethylaniline, p-toluidine, n-ethyl, n-ethyl-4-methyl-aniline, n-athyl-p-toluidin, 4-ethylamino toluene, ethyl 4-methylphenyl amine
N-ethyl-4-methylaniline
C9H13N
AASABFUMCBTXRL-UHFFFAOYSA-N
61164
CCNC1=CC=C(C=C1)C

Specifications

N-Ethyl-p-toluidine
0.9380g/mL
89°C
Glass bottle
CH3C6H4NHC2H5
0.938
AASABFUMCBTXRL-UHFFFAOYSA-N
N-ethyl-4-methylaniline
61164
97%
622-57-1
218°C
96% min. (GC)
C9H13N
100g
n-ethyl-p-toluidine, benzenamine, n-ethyl-4-methyl, n-ethyl-4-toluidine, n-ethyl-p-methylaniline, p-methyl-n-ethylaniline, p-toluidine, n-ethyl, n-ethyl-4-methyl-aniline, n-athyl-p-toluidin, 4-ethylamino toluene, ethyl 4-methylphenyl amine
CCNC1=CC=C(C=C1)C
135.21
135.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

GHS Signal Word: Danger

EINECSNumber : 210-742-9

SDS
Documents

Documents

Promotions

Promotions