missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-p-toluidine, 97%, Thermo Scientific™

Catalog No. AC450780250
Change view
Click to view available options
Quantity:
25g
100g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450780250 25g
AC450781000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450780250 Supplier Thermo Scientific Chemicals Supplier No. 450780250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 622-57-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
InChI Key AASABFUMCBTXRL-UHFFFAOYSA-N
Synonym n-ethyl-p-toluidine, benzenamine, n-ethyl-4-methyl, n-ethyl-4-toluidine, n-ethyl-p-methylaniline, p-methyl-n-ethylaniline, p-toluidine, n-ethyl, n-ethyl-4-methyl-aniline, n-athyl-p-toluidin, 4-ethylamino toluene, ethyl 4-methylphenyl amine
PubChem CID 61164
IUPAC Name N-ethyl-4-methylaniline
SMILES CCNC1=CC=C(C=C1)C

Specifications

Density 0.9380g/mL
Boiling Point 218°C
Flash Point 89°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4NHC2H5
Quantity 25g
Specific Gravity 0.938
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material N-Ethyl-p-toluidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
EINECSNumber 210-742-9

RUO – Research Use Only