missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Ethyl-p-toluenesulfonamide 98.0+%, TCI America™
$41.02 - $41.02
Chemical Identifiers
| CAS | 80-39-7 |
|---|---|
| Molecular Formula | C9H13NO2S |
| Molecular Weight (g/mol) | 199.268 |
| InChI Key | OHPZPBNDOVQJMH-UHFFFAOYSA-N |
| Synonym | n-ethyl-p-toluenesulfonamide, santicizer 3, n-tosylethylamine, benzenesulfonamide, n-ethyl-4-methyl, n-ethyl-p-tolylsulfonamide, n-ethyl-4-toluenesulfonamide, p-tolueneethylsulfonamide, ethyl tosylamide, p-toluenesulfonyl-n-ethylamide, n-ethyl-p-methylbenzenesulfonamide |
| PubChem CID | 6637 |
| IUPAC Name | N-ethyl-4-methylbenzenesulfonamide |
| SMILES | CCNS(=O)(=O)C1=CC=C(C=C1)C |
Chemical Identifiers
| 80-39-7 | |
| 199.268 | |
| n-ethyl-p-toluenesulfonamide, santicizer 3, n-tosylethylamine, benzenesulfonamide, n-ethyl-4-methyl, n-ethyl-p-tolylsulfonamide, n-ethyl-4-toluenesulfonamide, p-tolueneethylsulfonamide, ethyl tosylamide, p-toluenesulfonyl-n-ethylamide, n-ethyl-p-methylbenzenesulfonamide | |
| N-ethyl-4-methylbenzenesulfonamide |
| C9H13NO2S | |
| OHPZPBNDOVQJMH-UHFFFAOYSA-N | |
| 6637 | |
| CCNS(=O)(=O)C1=CC=C(C=C1)C |
Specifications
| 80-39-7 | |
| White | |
| C9H13NO2S | |
| n-ethyl-p-toluenesulfonamide, santicizer 3, n-tosylethylamine, benzenesulfonamide, n-ethyl-4-methyl, n-ethyl-p-tolylsulfonamide, n-ethyl-4-toluenesulfonamide, p-tolueneethylsulfonamide, ethyl tosylamide, p-toluenesulfonyl-n-ethylamide, n-ethyl-p-methylbenzenesulfonamide | |
| CCNS(=O)(=O)C1=CC=C(C=C1)C | |
| 199.268 | |
| 199.27 | |
| Crystalline Powder |
| 65°C | |
| 208°C | |
| 25 g | |
| OHPZPBNDOVQJMH-UHFFFAOYSA-N | |
| N-ethyl-4-methylbenzenesulfonamide | |
| 6637 | |
| ≥98.0% (HPLC,N) | |
| N-Ethyl-p-toluenesulfonamide |
Safety and Handling
EINECSNumber : (3)-1929
TSCA : Yes