missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-m-toluidine 98.0+%, TCI America™

Catalog No. p-7128880
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E017825ML 25 mL
E0178500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E017825ML Supplier TCI America Supplier No. E017825ML
Only null left

Chemical Identifiers

CAS 102-27-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00009027
InChI Key GUYMMHOQXYZMJQ-UHFFFAOYSA-N
Synonym n-ethyl-m-toluidine, benzenamine, n-ethyl-3-methyl, m-toluidine, n-ethyl, m-methyl-n-ethylaniline, n-ethyl-3-methylbenzenamine, unii-12k193mtcr, 3-ethylamino toluene, n-ethyl-meta-toluidine, n-ethyl-3-methyl-aniline, toluene, 3-ethylamino
PubChem CID 7603
IUPAC Name N-ethyl-3-methylaniline
SMILES CCNC1=CC=CC(C)=C1

Specifications

Boiling Point 221°C
Quantity 25 mL
UN Number 2754
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Ethyl-m-toluidine
EINECSNumber (3)-0187&(3)-0193
RTECSNumber CY0440000
TSCA Yes