missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Ethyl-m-toluidine 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 221°C |
| Quantity | 25 mL |
| UN Number | 2754 |
| Formula Weight | 135.21 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | N-Ethyl-m-toluidine |
Safety and Handling
| EINECSNumber | (3)-0187&(3)-0193 |
| RTECSNumber | CY0440000 |
| TSCA | Yes |