missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-4-chloroaniline 98.0+%, TCI America™

Catalog No. e042025g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E042025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E042025G Supplier TCI America Supplier No. E042025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13519-75-0
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00018594
InChI Key KLWDPIXDUVYHMS-UHFFFAOYSA-N
Synonym 4-Chloro-N-ethylaniline
PubChem CID 587841
IUPAC Name 4-chloro-N-ethylaniline
SMILES CCNC1=CC=C(C=C1)Cl

Specifications

Boiling Point 101°C
Quantity 25 g
Formula Weight 155.63
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Ethyl-4-chloroaniline