accessibility menu, dialog, popup

UserName

CH3NHCl

CAS RN 13519-75-0

IUPAC Name: 4-chloro-N-ethylaniline
Synonyms: 4-chloro-N-ethylaniline
Molecular Weight (g/mol): 155.63
Molecular Formula: C8H10ClN
InChi Key: KLWDPIXDUVYHMS-UHFFFAOYSA-N
SMILES: CCNC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results