missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Ethoxycarbonyl)nortropinone 98.0+%, TCI America™
Supplier: TCI America E083425G
Specifications
| N-(Ethoxycarbonyl)nortropinone | |
| Yellow | |
| C10H15NO3 | |
| 25 g | |
| ANEJUHJDPGTVIO-UHFFFAOYSA-N | |
| ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate | |
| 401882 | |
| ≥98.0% (GC) |
| 32499-64-2 | |
| 153°C | |
| MFCD00078171 | |
| ethyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-ethoxycarbonyl nortropinone, n-carbethoxy-4-nortropinone, n-carbethoxy-4-tropinone, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one, 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid ethyl ester, 8-azabicyclo 3.2.1 octane-8-carboxylic acid, 3-oxo-, ethyl ester, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, n-carbethoxynortropinone | |
| CCOC(=O)N1C2CCC1CC(=O)C2 | |
| 197.234 | |
| 197.23 | |
| Liquid |
Chemical Identifiers
| 32499-64-2 | |
| 197.234 | |
| ANEJUHJDPGTVIO-UHFFFAOYSA-N | |
| 401882 | |
| CCOC(=O)N1C2CCC1CC(=O)C2 |
| C10H15NO3 | |
| MFCD00078171 | |
| ethyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-ethoxycarbonyl nortropinone, n-carbethoxy-4-nortropinone, n-carbethoxy-4-tropinone, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one, 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid ethyl ester, 8-azabicyclo 3.2.1 octane-8-carboxylic acid, 3-oxo-, ethyl ester, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, n-carbethoxynortropinone | |
| ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate |
Safety and Handling
TSCA : No