missing translation for 'onlineSavingsMsg'
Learn More

N-(Ethoxycarbonyl)nortropinone 98.0+%, TCI America™

Catalog No. p-7129175
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
E08345G 5 g
E083425G 25 g
2 options

Catalog No. E08345G

Supplier: TCI America E08345G

Only null left

Chemical Identifiers

CAS 32499-64-2
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.234
MDL Number MFCD00078171
InChI Key ANEJUHJDPGTVIO-UHFFFAOYSA-N
Synonym ethyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-ethoxycarbonyl nortropinone, n-carbethoxy-4-nortropinone, n-carbethoxy-4-tropinone, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one, 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid ethyl ester, 8-azabicyclo 3.2.1 octane-8-carboxylic acid, 3-oxo-, ethyl ester, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, n-carbethoxynortropinone
PubChem CID 401882
IUPAC Name ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
SMILES CCOC(=O)N1C2CCC1CC(=O)C2

Specifications

Color Yellow
Boiling Point 153°C
Quantity 5 g
Formula Weight 197.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(Ethoxycarbonyl)nortropinone