missing translation for 'onlineSavingsMsg'
Learn More

N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%

Catalog No. p-3736425
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC165350250 25 g Glass bottle
AC165351000 100 g Glass bottle
2 options

Catalog No. AC165350250

Supplier: Thermo Scientific Chemicals 165350250

Only null left

CAS: 16357-59-8 | C14H17NO3 | 247.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 16357-59-8
Molecular Formula C14H17NO3
Molecular Weight (g/mol) 247.29
MDL Number MFCD00006703
InChI Key GKQLYSROISKDLL-UHFFFAOYNA-N
Synonym eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate
PubChem CID 27833
IUPAC Name ethyl 2-ethoxy-2H-quinoline-1-carboxylate
SMILES CCOC1C=CC2=CC=CC=C2N1C(=O)OCC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 63.0°C to 66.0°C
Color White to Beige
Boiling Point 125.0°C to 128.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, V,3, 28
Fieser 02,191; 03,137; 04,223
Merck Index 15, 3566
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 247.29
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material N-Ethoxycarbonyl-2-ethoxy-1, 2-dihydroquinoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
EINECSNumber 240-418-2

RUO – Research Use Only