missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(epsilon)-Acetyl-L-lysine, 99%

Catalog No. p-7042879
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6413903 1 g
AAJ6413906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6413903 Supplier Thermo Scientific Chemicals Supplier No. J6413903

CAS: 692-04-6 | C8H16N2O3 | 188.23 g/mol

N(epsilon)-Acetyl-L-lysine is an R-chain N-acetylated α amino acid used together with other lysine analogues to differentiate and characterized various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N(epsilon)-Acetyl-L-lysine is an R-chain N-acetylated α amino acid used together with other lysine analogues to differentiate and characterized various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.

Solubility
Soluble in water (miscible), and 80% acetic acid (50 mg/ml).

Notes
Hygroscopic. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 692-04-6
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD00002639
InChI Key DTERQYGMUDWYAZ-ZETCQYMHSA-N
PubChem CID 92832
ChEBI CHEBI:17752
IUPAC Name (2S)-6-acetamido-2-aminohexanoic acid
SMILES CC(=O)NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 250°C (decomposition)
Color White
Synonym n-epsilon-acetyl-l-lysine, n6-acetyl-l-lysine, nepsilon-acetyl-l-lysine, n-epsilon-acetyllysine, h-lys ac-oh, n6-acetyllysine, s-6-acetamido-2-aminohexanoic acid, n 6-acetyl-l-lysine, n 6-acetyllysine, n epsilon-acetyl-l-lysine
Solubility Information Soluble in water (miscible),and 80% acetic acid (50mg/ml).
Quantity 1 g
Formula Weight 188.2
Percent Purity 99%
Sensitivity Hygroscopic
Physical Form Powder
Chemical Name or Material N(epsilon)-Acetyl-L-lysine
EINECSNumber 211-725-9
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only