missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Dodecyltrimethylammonium bromide, MilliporeSigma™
For synthesis
Supplier: Technidata 8410220100
Specifications
| n-Dodecyltrimethylammonium bromide | |
| 237°C to 245°C | |
| Odorless | |
| 100 g | |
| XJWSAJYUBXQQDR-UHFFFAOYSA-M | |
| dodecyl(trimethyl)azanium;bromide | |
| 14249 | |
| 308.34 |
| 1119-94-4 | |
| White | |
| C15H34BrN | |
| dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide | |
| CCCCCCCCCCCC[N+](C)(C)C.[Br-] | |
| 308.348 | |
| CHEBI:282662 | |
| Solid |
Chemical Identifiers
| 1119-94-4 | |
| 308.348 | |
| dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide | |
| CHEBI:282662 | |
| CCCCCCCCCCCC[N+](C)(C)C.[Br-] |
| C15H34BrN | |
| XJWSAJYUBXQQDR-UHFFFAOYSA-M | |
| 14249 | |
| dodecyl(trimethyl)azanium;bromide |
Safety and Handling
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H301: Toxic if swallowed.
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 214-290-3
Recommended Storage : 15° to 25°C (59° to 77°F); Tightly Closed. Dry. Keep in well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons.