accessibility menu, dialog, popup

UserName

BrH3CNCH3CH3H3C

CAS RN 1119-94-4

IUPAC Name: dodecyltrimethylazanium bromide
Synonyms: DTAB laurtrimonium bromide morpan D LTAB dctab
Molecular Weight (g/mol): 308.35
Molecular Formula: C15H34BrN
InChi Key: XJWSAJYUBXQQDR-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (4)
  • (8)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (6)
Quantity 
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (4)
  • (2)
Grade 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (6)
  • (4)
  • (8)
  • (2)
  • (3)
  • (1)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)