missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Dodecyltrimethylammonium bromide, MilliporeSigma™
Specifications
Specifications
| Melting Point | 237°C to 245°C |
| Color | White |
| Odor | Odorless |
| Quantity | 25 g |
| Formula Weight | 308.34 |
| Percent Purity | ≥97.0% |
| Physical Form | Solid |
| Chemical Name or Material | n-Dodecyltrimethylammonium bromide |
Safety and Handling
| Hazard Category | H301: Toxic if swallowed. |
| Hazard Statement | H315: Causes skin irritation. |
| Precautionary Statement | H319: Causes serious eye irritation. |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 214-290-3 |
| Recommended Storage | 15° to 25°C (59° to 77°F); Tightly Closed. Dry. Keep in well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons. |