Learn More
n-Dodecylpyridinium chloride hydrate, 98%
CAS: 207234-02-4 | C17H30ClN | 283.88 g/mol
$107.52 - $268.33
Chemical Identifiers
| CAS | 207234-02-4 |
|---|---|
| Molecular Formula | C17H30ClN |
| Molecular Weight (g/mol) | 283.88 |
| MDL Number | MFCD00011987 |
| InChI Key | GKQHIYSTBXDYNQ-UHFFFAOYSA-M |
| Synonym | 1-dodecylpyridin-1-ium chloride hydrate, laurylpyridinum chloride, 1-dodecylpyridinium chloride hydrate, 1-dodecylpyridin-1-ium hydrate chloride, c17h30n.cl.h2o, n-dodecylpyridinium chloride hydrate, dodecylpyridinium chloride monohydrate, n-dodecylpyridinium chloride monohydrate, 1-dodecylpyridin-1-ium chloride-water 1/1/1 |
| PubChem CID | 16211802 |
| IUPAC Name | 1-dodecylpyridin-1-ium;chloride;hydrate |
| SMILES | [Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2349214
|
Thermo Scientific Chemicals
B2349214 |
25 g |
Each for $107.52
|
|
|||||
|
AAB2349222
|
Thermo Scientific Chemicals
B2349222 |
100 g |
Each for $268.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 207234-02-4 | |
| 283.88 | |
| GKQHIYSTBXDYNQ-UHFFFAOYSA-M | |
| 16211802 | |
| [Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1 |
| C17H30ClN | |
| MFCD00011987 | |
| 1-dodecylpyridin-1-ium chloride hydrate, laurylpyridinum chloride, 1-dodecylpyridinium chloride hydrate, 1-dodecylpyridin-1-ium hydrate chloride, c17h30n.cl.h2o, n-dodecylpyridinium chloride hydrate, dodecylpyridinium chloride monohydrate, n-dodecylpyridinium chloride monohydrate, 1-dodecylpyridin-1-ium chloride-water 1/1/1 | |
| 1-dodecylpyridin-1-ium;chloride;hydrate |
Specifications
| 207234-02-4 | |
| C17H30ClN | |
| 25 g | |
| 1-dodecylpyridin-1-ium chloride hydrate, laurylpyridinum chloride, 1-dodecylpyridinium chloride hydrate, 1-dodecylpyridin-1-ium hydrate chloride, c17h30n.cl.h2o, n-dodecylpyridinium chloride hydrate, dodecylpyridinium chloride monohydrate, n-dodecylpyridinium chloride monohydrate, 1-dodecylpyridin-1-ium chloride-water 1/1/1 | |
| [Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1 | |
| 283.88 | |
| 283.89 (Anhydrous) | |
| n-Dodecylpyridinium chloride hydrate |
| 88°C to 90°C | |
| MFCD00011987 | |
| Hygroscopic | |
| GKQHIYSTBXDYNQ-UHFFFAOYSA-M | |
| 1-dodecylpyridin-1-ium;chloride;hydrate | |
| 16211802 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 203-232-2
RTECSNumber : UU4017070
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only