missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecylpyridinium chloride hydrate, 98%

Catalog No. AAB2349214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2349214 25 g
AAB2349222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2349214 Supplier Thermo Scientific Chemicals Supplier No. B2349214
Only null left

CAS: 207234-02-4 | C17H30ClN | 283.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207234-02-4
Molecular Formula C17H30ClN
Molecular Weight (g/mol) 283.88
MDL Number MFCD00011987
InChI Key GKQHIYSTBXDYNQ-UHFFFAOYSA-M
Synonym 1-dodecylpyridin-1-ium chloride hydrate, laurylpyridinum chloride, 1-dodecylpyridinium chloride hydrate, 1-dodecylpyridin-1-ium hydrate chloride, c17h30n.cl.h2o, n-dodecylpyridinium chloride hydrate, dodecylpyridinium chloride monohydrate, n-dodecylpyridinium chloride monohydrate, 1-dodecylpyridin-1-ium chloride-water 1/1/1
PubChem CID 16211802
IUPAC Name 1-dodecylpyridin-1-ium;chloride;hydrate
SMILES [Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1

Specifications

Melting Point 88°C to 90°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 283.89 (Anhydrous)
Percent Purity 98%
Chemical Name or Material n-Dodecylpyridinium chloride hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-232-2
RTECSNumber UU4017070
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only