missing translation for 'onlineSavingsMsg'
Learn More

n-Docosane, 99%

Catalog No. p-3778485
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158780250 25 g Glass bottle
AC158781000 100 g Glass bottle
AC158785000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158780250 Supplier Thermo Scientific Chemicals Supplier No. 158780250
Only null left

CAS: 629-97-0 | C22H46 | 310.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-97-0
Molecular Formula C22H46
Molecular Weight (g/mol) 310.61
MDL Number MFCD00009348
InChI Key HOWGUJZVBDQJKV-UHFFFAOYSA-N
Synonym n-docosane, dokosan, unii-ow99q363ko, heneicosane, methyl, ch3-ch2 20-ch3, docosane, analytical standard, twt, normal-docosane, docosane, n, docosane
PubChem CID 12405
ChEBI CHEBI:46050
IUPAC Name docosane
SMILES CCCCCCCCCCCCCCCCCCCCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 43.0°C to 46.0°C
Color White
Density 0.7700g/mL
Boiling Point 369.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)20CH3
Quantity 25 g
Beilstein 01, 174
Specific Gravity 0.77
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene and toluene
Formula Weight 310.6
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material n-Docosane
Show More Show Less
EINECSNumber 211-121-5

RUO – Research Use Only