accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 629-97-0

IUPAC Name: docosane
Synonyms: docosane dokosan
Molecular Weight (g/mol): 310.61
Molecular Formula: C22H46
InChi Key: HOWGUJZVBDQJKV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCC
Boiling Point 
  • (2)
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (1)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (5)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)