missing translation for 'onlineSavingsMsg'
Learn More

n-Docosane, 99%

CAS: 629-97-0 | C22H46 | 310.61 g/mol

Supplier:  Thermo Scientific Chemicals A1805022

 View more versions of this product

Catalog No. AAA1805022

Only null left
Explore available promotions

Description

Description

It is employed as a perfuming agent in cosmetics industry. It is also used in organic synthesis, calibration, and temperature sensing equipment. Docosane is used to investigate commercially available waxes in the form of thin disc samples as possible diffraction intensity standards for macromolecular crystallography synchrotron beamlines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a perfuming agent in cosmetics industry. It is also used in organic synthesis, calibration, and temperature sensing equipment. Docosane is used to investigate commercially available waxes in the form of thin disc samples as possible diffraction intensity standards for macromolecular crystallography synchrotron beamlines.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

n-Docosane
43°C to 46°C
368°C to 369°C
C22H46
MFCD00009348
1702206
Insoluble in water. Soluble in alcohol.
CCCCCCCCCCCCCCCCCCCCCC
310.61
CHEBI:46050
99%
629-97-0
0.777
>110°C (230°F)
1.4455
100 g
n-docosane, dokosan, unii-ow99q363ko, heneicosane, methyl, ch3-ch2 20-ch3, docosane, analytical standard, twt, normal-docosane, docosane, n, docosane
HOWGUJZVBDQJKV-UHFFFAOYSA-N
docosane
12405
310.61

Chemical Identifiers

629-97-0
310.61
HOWGUJZVBDQJKV-UHFFFAOYSA-N
12405
docosane
C22H46
MFCD00009348
n-docosane, dokosan, unii-ow99q363ko, heneicosane, methyl, ch3-ch2 20-ch3, docosane, analytical standard, twt, normal-docosane, docosane, n, docosane
CHEBI:46050
CCCCCCCCCCCCCCCCCCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-121-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only