missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Diphenylmethylene)glycine tert-Butyl Ester 98.0+%, TCI America™
Supplier: TCI America D232225G
Specifications
| N-(Diphenylmethylene)glycine tert-Butyl Ester | |
| 116°C | |
| C19H21NO2 | |
| 25 g | |
| YSHDPXQDVKNPKA-UHFFFAOYSA-N | |
| tert-butyl 2-(benzhydrylideneamino)acetate | |
| 688171 | |
| ≥98.0% (HPLC,N) |
| 81477-94-3 | |
| White | |
| MFCD00134280 | |
| n-diphenylmethylene glycine tert-butyl ester, diphenylmethylene-glycine t-butyl ester, tert-butyl 2-diphenylmethylene amino acetate, n-diphenylmethylene glycerine tert-butyl ester, tert-butyl 2-diphenylmethyleneamino acetate, diphenylmethyleneamino acetic acid tert-butyl ester, tert-butyl diphenylmethyleneamino acetate, n-diphenylmethylene glycinetert-butylester, tert-butyl 2-benzhydrylideneamino acetate, glycine, n-diphenylmethylene-, 1,1-dimethylethyl ester | |
| CC(C)(C)OC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2 | |
| 295.382 | |
| 295.38 | |
| Crystalline Powder |
Chemical Identifiers
| 81477-94-3 | |
| 295.382 | |
| YSHDPXQDVKNPKA-UHFFFAOYSA-N | |
| 688171 | |
| CC(C)(C)OC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2 |
| C19H21NO2 | |
| MFCD00134280 | |
| n-diphenylmethylene glycine tert-butyl ester, diphenylmethylene-glycine t-butyl ester, tert-butyl 2-diphenylmethylene amino acetate, n-diphenylmethylene glycerine tert-butyl ester, tert-butyl 2-diphenylmethyleneamino acetate, diphenylmethyleneamino acetic acid tert-butyl ester, tert-butyl diphenylmethyleneamino acetate, n-diphenylmethylene glycinetert-butylester, tert-butyl 2-benzhydrylideneamino acetate, glycine, n-diphenylmethylene-, 1,1-dimethylethyl ester | |
| tert-butyl 2-(benzhydrylideneamino)acetate |
Safety and Handling
TSCA : No