missing translation for 'onlineSavingsMsg'
Learn More

N-(Diphenylmethylene)glycine tert-Butyl Ester 98.0+%, TCI America™

Catalog No. D23225G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23225G 5 g
D23221G 1 g
D232225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D23225G Supplier TCI America Supplier No. D23225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81477-94-3
Molecular Formula C19H21NO2
Molecular Weight (g/mol) 295.382
MDL Number MFCD00134280
InChI Key YSHDPXQDVKNPKA-UHFFFAOYSA-N
Synonym n-diphenylmethylene glycine tert-butyl ester, diphenylmethylene-glycine t-butyl ester, tert-butyl 2-diphenylmethylene amino acetate, n-diphenylmethylene glycerine tert-butyl ester, tert-butyl 2-diphenylmethyleneamino acetate, diphenylmethyleneamino acetic acid tert-butyl ester, tert-butyl diphenylmethyleneamino acetate, n-diphenylmethylene glycinetert-butylester, tert-butyl 2-benzhydrylideneamino acetate, glycine, n-diphenylmethylene-, 1,1-dimethylethyl ester
PubChem CID 688171
IUPAC Name tert-butyl 2-(benzhydrylideneamino)acetate
SMILES CC(C)(C)OC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 116°C
Color White
Quantity 5 g
Formula Weight 295.38
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(Diphenylmethylene)glycine tert-Butyl Ester