Learn More
N-(Diphenylmethylene)glycine tert-butyl ester, 99%
CAS: 81477-94-3 | C19H21NO2 | 295.382 g/mol
$162.58 - $522.53
Chemical Identifiers
| CAS | 81477-94-3 |
|---|---|
| Molecular Formula | C19H21NO2 |
| Molecular Weight (g/mol) | 295.382 |
| MDL Number | MFCD00134280 |
| InChI Key | YSHDPXQDVKNPKA-UHFFFAOYSA-N |
| Synonym | n-diphenylmethylene glycine tert-butyl ester, diphenylmethylene-glycine t-butyl ester, tert-butyl 2-diphenylmethylene amino acetate, n-diphenylmethylene glycerine tert-butyl ester, tert-butyl 2-diphenylmethyleneamino acetate, diphenylmethyleneamino acetic acid tert-butyl ester, tert-butyl diphenylmethyleneamino acetate, n-diphenylmethylene glycinetert-butylester, tert-butyl 2-benzhydrylideneamino acetate, glycine, n-diphenylmethylene-, 1,1-dimethylethyl ester |
| PubChem CID | 688171 |
| IUPAC Name | tert-butyl 2-(benzhydrylideneamino)acetate |
| SMILES | CC(C)(C)OC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3270903
|
Thermo Scientific Chemicals
H3270903 |
1 g |
Each for $162.58
|
|
|||||
|
AAH3270906
|
Thermo Scientific Chemicals
H3270906 |
5 g |
Each for $522.53
|
|
|||||
Description
N-(Diphenylmethylene)glycine tert-butyl ester is a useful dye for biological research purposes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-(Diphenylmethylene)glycine tert-butyl ester is a useful dye for biological research purposes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 81477-94-3 | |
| 295.382 | |
| YSHDPXQDVKNPKA-UHFFFAOYSA-N | |
| 688171 | |
| CC(C)(C)OC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2 |
| C19H21NO2 | |
| MFCD00134280 | |
| n-diphenylmethylene glycine tert-butyl ester, diphenylmethylene-glycine t-butyl ester, tert-butyl 2-diphenylmethylene amino acetate, n-diphenylmethylene glycerine tert-butyl ester, tert-butyl 2-diphenylmethyleneamino acetate, diphenylmethyleneamino acetic acid tert-butyl ester, tert-butyl diphenylmethyleneamino acetate, n-diphenylmethylene glycinetert-butylester, tert-butyl 2-benzhydrylideneamino acetate, glycine, n-diphenylmethylene-, 1,1-dimethylethyl ester | |
| tert-butyl 2-(benzhydrylideneamino)acetate |
Specifications
| 81477-94-3 | |
| C19H21NO2 | |
| MFCD00134280 | |
| n-diphenylmethylene glycine tert-butyl ester, diphenylmethylene-glycine t-butyl ester, tert-butyl 2-diphenylmethylene amino acetate, n-diphenylmethylene glycerine tert-butyl ester, tert-butyl 2-diphenylmethyleneamino acetate, diphenylmethyleneamino acetic acid tert-butyl ester, tert-butyl diphenylmethyleneamino acetate, n-diphenylmethylene glycinetert-butylester, tert-butyl 2-benzhydrylideneamino acetate, glycine, n-diphenylmethylene-, 1,1-dimethylethyl ester | |
| YSHDPXQDVKNPKA-UHFFFAOYSA-N | |
| tert-butyl 2-(benzhydrylideneamino)acetate | |
| 688171 | |
| 99% |
| 113°C to 115°C | |
| (C6H5)2C=NCH2COOC(CH3)3 | |
| 1 g | |
| Insoluble in water. | |
| CC(C)(C)OC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2 | |
| 295.382 | |
| 295.38 | |
| N-(Diphenylmethylene)glycine tert-butyl ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only