missing translation for 'onlineSavingsMsg'
Learn More

N-(Diphenylmethylene)glycine tert-butyl ester, 99%

Catalog No. AAH3270903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3270903 1 g
AAH3270906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3270903 Supplier Thermo Scientific Chemicals Supplier No. H3270903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 81477-94-3 | C19H21NO2 | 295.382 g/mol

N-(Diphenylmethylene)glycine tert-butyl ester is a useful dye for biological research purposes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Diphenylmethylene)glycine tert-butyl ester is a useful dye for biological research purposes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 81477-94-3
Molecular Formula C19H21NO2
Molecular Weight (g/mol) 295.382
MDL Number MFCD00134280
InChI Key YSHDPXQDVKNPKA-UHFFFAOYSA-N
Synonym n-diphenylmethylene glycine tert-butyl ester, diphenylmethylene-glycine t-butyl ester, tert-butyl 2-diphenylmethylene amino acetate, n-diphenylmethylene glycerine tert-butyl ester, tert-butyl 2-diphenylmethyleneamino acetate, diphenylmethyleneamino acetic acid tert-butyl ester, tert-butyl diphenylmethyleneamino acetate, n-diphenylmethylene glycinetert-butylester, tert-butyl 2-benzhydrylideneamino acetate, glycine, n-diphenylmethylene-, 1,1-dimethylethyl ester
PubChem CID 688171
IUPAC Name tert-butyl 2-(benzhydrylideneamino)acetate
SMILES CC(C)(C)OC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 113°C to 115°C
Linear Formula (C6H5)2C=NCH2COOC(CH3)3
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 295.38
Percent Purity 99%
Chemical Name or Material N-(Diphenylmethylene)glycine tert-butyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only