missing translation for 'onlineSavingsMsg'
Learn More

N-(Diphenylmethylene)glycine Ethyl Ester 97.0+%, TCI America™

Catalog No. p-7127945
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38335G 5 g
D383325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38335G Supplier TCI America Supplier No. D38335G
Only null left

Chemical Identifiers

CAS 69555-14-2
Molecular Formula C17H17NO2
Molecular Weight (g/mol) 267.328
MDL Number MFCD00010590
InChI Key QUGJYNGNUBHTNS-UHFFFAOYSA-N
Synonym ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylidene amino acetate, diphenylmethylene glycine ethylester, ethyl diphenylmethylenamino acetate, diphenylmethylene glycine ethyl ester
PubChem CID 319508
IUPAC Name ethyl 2-(benzhydrylideneamino)acetate
SMILES CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 195°C
Quantity 5 g
Formula Weight 267.33
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(Diphenylmethylene)glycine Ethyl Ester