missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Diphenylmethylene)aminoacetonitrile, 99%
CAS: 70591-20-7 | C15H12N2 | 220.28 g/mol
$152.42 - $152.42
Chemical Identifiers
| CAS | 70591-20-7 |
|---|---|
| Molecular Formula | C15H12N2 |
| Molecular Weight (g/mol) | 220.28 |
| MDL Number | MFCD00009970 |
| InChI Key | VRLJFRODHVSTIK-UHFFFAOYSA-N |
| Synonym | n-diphenylmethylene aminoacetonitrile, 2-diphenylmethylene amino acetonitrile, benzhydrylidene-amino-acetonitrile, 2-benzhydrylideneamino acetonitrile, 2-diphenylmethylidene amino acetonitrile, acetonitrile, diphenylmethylene amino, 4,4-diphenyl-3-azabut-3-enenitrile, pubchem13883, benzhydrylideneaminoacetonitrile |
| PubChem CID | 612957 |
| IUPAC Name | 2-(benzhydrylideneamino)acetonitrile |
| SMILES | N#CCN=C(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC124140050
|
Thermo Scientific Chemicals
124140050 |
5 g | Glass bottle |
Each for $152.42
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 70591-20-7 | |
| 220.28 | |
| VRLJFRODHVSTIK-UHFFFAOYSA-N | |
| 612957 | |
| N#CCN=C(C1=CC=CC=C1)C1=CC=CC=C1 |
| C15H12N2 | |
| MFCD00009970 | |
| n-diphenylmethylene aminoacetonitrile, 2-diphenylmethylene amino acetonitrile, benzhydrylidene-amino-acetonitrile, 2-benzhydrylideneamino acetonitrile, 2-diphenylmethylidene amino acetonitrile, acetonitrile, diphenylmethylene amino, 4,4-diphenyl-3-azabut-3-enenitrile, pubchem13883, benzhydrylideneaminoacetonitrile | |
| 2-(benzhydrylideneamino)acetonitrile |
Specifications
| 70591-20-7 | |
| 100.0 | |
| Gray or White to Yellow | |
| 98.5% min. (HPLC) | |
| C15H12N2 | |
| MFCD00009970 | |
| n-diphenylmethylene aminoacetonitrile, 2-diphenylmethylene amino acetonitrile, benzhydrylidene-amino-acetonitrile, 2-benzhydrylideneamino acetonitrile, 2-diphenylmethylidene amino acetonitrile, acetonitrile, diphenylmethylene amino, 4,4-diphenyl-3-azabut-3-enenitrile, pubchem13883, benzhydrylideneaminoacetonitrile | |
| N#CCN=C(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 220.28 | |
| 220.27 | |
| Crystals or Powder |
| 98.5 | |
| 83.0°C to 86.0°C | |
| Authentic | |
| Glass bottle | |
| (C6H5)2C=NCH2CN | |
| 5 g | |
| VRLJFRODHVSTIK-UHFFFAOYSA-N | |
| 2-(benzhydrylideneamino)acetonitrile | |
| 612957 | |
| 99% | |
| N-(Diphenylmethylene)aminoacetonitrile |
RUO – Research Use Only