missing translation for 'onlineSavingsMsg'
Learn More

n-Decyldimethylchlorosilane, 97%, Thermo Scientific™

$434.26 - $434.26

Chemical Identifiers

CAS 38051-57-9
Molecular Formula C12H27ClSi
Molecular Weight (g/mol) 234.883
MDL Number MFCD00054168
InChI Key ZLZGHBNDPINFKG-UHFFFAOYSA-N
Synonym chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa
PubChem CID 2758020
IUPAC Name chloro-decyl-dimethylsilane
SMILES CCCCCCCCCC[Si](C)(C)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4276918
View Documents
Thermo Scientific Chemicals
4276918
50 g
Each for $434.26
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

38051-57-9
234.883
ZLZGHBNDPINFKG-UHFFFAOYSA-N
2758020
CCCCCCCCCC[Si](C)(C)Cl
C12H27ClSi
MFCD00054168
chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa
chloro-decyl-dimethylsilane

Specifications

98°C (2mmHg)
Liquid
C12H27ClSi
UN1760
chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa
CCCCCCCCCC[Si](C)(C)Cl
234.883
234.88
1.441
0.866 g/mL
38051-57-9
50 g
MFCD00054168
137°C (279°F)
ZLZGHBNDPINFKG-UHFFFAOYSA-N
chloro-decyl-dimethylsilane
2758020
97%
Moisture sensitive
n-Decyldimethylchlorosilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 253-761-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'