accessibility menu, dialog, popup

UserName

H3CSiCH3Cl

CAS RN 38051-57-9

IUPAC Name: (10-chlorodecyl)dimethylsilyl
Synonyms: (10-chlorodecyl)dimethylsilyl
Molecular Weight (g/mol): 233.88
Molecular Formula: C12H26ClSi
InChi Key: RXEZDPFAWKPHOG-UHFFFAOYSA-N
SMILES: C[Si](C)CCCCCCCCCCCl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)