missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclopropyl-4-nitrobenzenesulfonamide, 97%, Thermo Scientific™

Catalog No. AAH58673ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58673ME 500 mg
AAH58673MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH58673ME Supplier Thermo Scientific Chemicals Supplier No. H58673ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 549476-61-1
Molecular Formula C9H10N2O4S
Molecular Weight (g/mol) 242.249
MDL Number MFCD00458256
InChI Key SFXOKDPLGLHGIX-UHFFFAOYSA-N
PubChem CID 3434716
IUPAC Name N-cyclopropyl-4-nitrobenzenesulfonamide
SMILES C1CC1NS(=O)(=O)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Quantity 500 mg
Synonym n-cyclopropyl 4-nitrophenylsulfonamide, cyclopropyl 4-nitrophenyl sulfonyl amine, n-cyclopropyl4-nitrophenylsulfonamide, pubchem18638, acmc-209lkb, n-cyclopropyl-4-nitro-benzenesulfonamide, n-cyclopropyl-4-nitrobenzene-1-sulfonamide
Formula Weight 242.25
Percent Purity 97%
Chemical Name or Material N-Cyclopropyl-4-nitrobenzenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures