missing translation for 'onlineSavingsMsg'
Learn More

N-(Cyclopentyloxycarbonyloxy)succinimide 98.0+%, TCI America™

Catalog No. C269525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C269525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C269525G Supplier TCI America Supplier No. C269525G
Only null left

Chemical Identifiers

CAS 128595-07-3
Molecular Formula C10H13NO5
Molecular Weight (g/mol) 227.216
MDL Number MFCD04038113
InChI Key MKHDXOXWZKDUKB-UHFFFAOYSA-N
Synonym Cyclopentyl N-Succinimidyl Carbonate
PubChem CID 15927743
IUPAC Name cyclopentyl (2,5-dioxopyrrolidin-1-yl) carbonate
SMILES C1CCC(C1)OC(=O)ON2C(=O)CCC2=O

Specifications

Melting Point 80°C
Color White
Quantity 25 g
Formula Weight 227.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(Cyclopentyloxycarbonyloxy)succinimide
TSCA No
Recommended Storage Refrigerator