accessibility menu, dialog, popup

UserName

OOONOO

CAS RN 128595-07-3

IUPAC Name: cyclopentyl 2,5-dioxopyrrolidin-1-yl carbonate
Synonyms: cyclopentyl 2,5-dioxopyrrolidin-1-yl carbonate
Molecular Weight (g/mol): 227.22
Molecular Formula: C10H13NO5
InChi Key: MKHDXOXWZKDUKB-UHFFFAOYSA-N
SMILES: O=C(OC1CCCC1)ON1C(=O)CCC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results