missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclohexylsulfamic Acid 98.0+%, TCI America™

Catalog No. C0486500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0486500G 500 g
C048625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0486500G Supplier TCI America Supplier No. C0486500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-88-9
Molecular Formula C6H13NO3S
Molecular Weight (g/mol) 179.23
MDL Number MFCD00065234
InChI Key HCAJEUSONLESMK-UHFFFAOYSA-N
Synonym Cyclohexanesulfamic Acid
PubChem CID 7533
ChEBI CHEBI:15964
IUPAC Name N-cyclohexylsulfamic acid
SMILES OS(=O)(=O)NC1CCCCC1

Specifications

Melting Point 169°C
Color White
Quantity 500 g
Formula Weight 179.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Cyclohexylsulfamic Acid
EINECSNumber (3)-2480
RTECSNumber GV6950000
TSCA Yes