accessibility menu, dialog, popup

UserName

CAS RN 100-88-9

IUPAC Name: N-cyclohexylsulfamic acid
Synonyms: cyclamate sucaryl cyclohexylamidosulphuric acid polycat 200 cyclohexylamide sulfate 环拉酸 cyclohexylamidosulfuric acid cylamic acid
Molecular Weight (g/mol): 179.23
Molecular Formula: C6H13NO3S
InChi Key: HCAJEUSONLESMK-UHFFFAOYSA-N
SMILES: OS(=O)(=O)NC1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results