missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclohexyl-p-toluenesulfonamide 99.0+%, TCI America™

Catalog No. C203225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C203225G 25 g
C2032500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C203225G Supplier TCI America Supplier No. C203225G

Chemical Identifiers

CAS 80-30-8
Molecular Formula C13H19NO2S
Molecular Weight (g/mol) 253.36
MDL Number MFCD00014285
InChI Key DKYVVNLWACXMDW-UHFFFAOYSA-N
Synonym n-cyclohexyl-p-toluenesulfonamide, santicizer 1h, p-toluenesulfonamide, n-cyclohexyl, benzenesulfonamide, n-cyclohexyl-4-methyl, tosylamino cyclohexane, ccris 6008, unii-0qpv1ae9em, n-cyclohexyltoluene-4-sulphonamide, n-cyclohexyl-4-methyl-benzenesulfonamide, 0qpv1ae9em
PubChem CID 6633
IUPAC Name N-cyclohexyl-4-methylbenzene-1-sulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC1CCCCC1

Specifications

Melting Point 87°C
Color White
Boiling Point 350°C
Quantity 25 g
Formula Weight 253.36
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Cyclohexyl-p-toluenesulfonamide
EINECSNumber (3)-2481
RTECSNumber XT5617000
TSCA Yes