missing translation for 'onlineSavingsMsg'
Learn More

N-Cycloheptylmethylamine, 97%, Thermo Scientific Chemicals

Catalog No. p-7056875
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6604403 1 g
AAH6604406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6604403 Supplier Thermo Scientific Chemicals Supplier No. H6604403

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42870-65-5
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD08271751
InChI Key LRXSDHDEISIWQB-UHFFFAOYSA-N
Synonym n-cycloheptyl-n-methylamine, hexamethylenemethylamine, cycloheptanemethanamine, n-methylcycloheptylamine, cycloheptyl methyl amine, cycloheptyl-methyl-amine, n-methyl-n-cycloheptylamine, cycloheptanamine, n-methyl
PubChem CID 276275
IUPAC Name N-methylcycloheptanamine
SMILES CNC1CCCCCC1

Specifications

Quantity 1 g
Beilstein 2715358
Formula Weight 127.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Cycloheptylmethylamine
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only