accessibility menu, dialog, popup

UserName

CAS RN 42870-65-5

IUPAC Name: N-methylcycloheptanamine
Synonyms: N-methylcycloheptanamine
Molecular Weight (g/mol): 127.23
Molecular Formula: C8H17N
InChi Key: LRXSDHDEISIWQB-UHFFFAOYSA-N
SMILES: CNC1CCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results