missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cyanomethyl-N-methyl-4-nitroaniline 98.0+%, TCI America™
Supplier: TCI America C13495G
Specifications
N-Cyanomethyl-N-methyl-4-nitroaniline | |
Yellow | |
MFCD00191337 | |
3439 | |
CN(CC#N)C1=CC=C(C=C1)[N+](=O)[O-] | |
191.19 | |
191.19 | |
Crystalline Powder |
107023-66-5 | |
C9H9N3O2 | |
5 g | |
DJOYTAUERRJRAT-UHFFFAOYSA-N | |
2-(N-methyl-4-nitroanilino)acetonitrile | |
19889016 | |
≥98.0% (GC) |
Chemical Identifiers
107023-66-5 | |
191.19 | |
DJOYTAUERRJRAT-UHFFFAOYSA-N | |
2-(N-methyl-4-nitroanilino)acetonitrile |
C9H9N3O2 | |
MFCD00191337 | |
19889016 | |
CN(CC#N)C1=CC=C(C=C1)[N+](=O)[O-] |
Safety and Handling
TSCA : No