missing translation for 'onlineSavingsMsg'
Learn More

N-Chlorosaccharin 96.0+%, TCI America™

Catalog No. p-7125120
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16745G 5 g
C167425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16745G Supplier TCI America Supplier No. C16745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14070-51-0
Molecular Formula C7H4ClNO3S
Molecular Weight (g/mol) 217.623
MDL Number MFCD00274277
InChI Key VKWMGUNWDFIWNW-UHFFFAOYSA-N
Synonym n-chlorosaccharin, unii-g35nk0ctff, 2-chlorobenzo d isothiazol-3 2h-one 1,1-dioxide, g35nk0ctff, 1,2-benzisothiazol-3 2h-one, 2-chloro-, 1,1-dioxide, n-chlorosaccharine, ncsa, ncsac, n-chlorosaccharin mi, 2-chloro-1??,2-benzothiazole-1,1,3-trione
PubChem CID 4586058
IUPAC Name 2-chloro-1,1-dioxo-1,2-benzothiazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)N(S2(=O)=O)Cl

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 5 g
Formula Weight 217.62
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chlorosaccharin
TSCA No
Recommended Storage Refrigerator