missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Chloromethyl)phthalimide, 97%
Description
It is used as an agrochemical and medicine intermediates and is used in organic synthesis. Microporous membranes prepared from chemically modified polysulfone, which is a derivative of this compound have better streaming potential.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an agrochemical and medicine intermediates and is used in organic synthesis. Microporous membranes prepared from chemically modified polysulfone, which is a derivative of this compound have better streaming potential.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.
It is used as an agrochemical and medicine intermediates and is used in organic synthesis. Microporous membranes prepared from chemically modified polysulfone, which is a derivative of this compound have better streaming potential.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.
Specifications
Specifications
| Melting Point | 131°C to 135°C |
| Quantity | 25 g |
| Beilstein | 140942 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 195.61 |
| Percent Purity | 97% |
| Chemical Name or Material | N- (Chloromethyl)phthalimide |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 241-541-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only