accessibility menu, dialog, popup

UserName

CAS RN 17564-64-6

IUPAC Name: 2-(chloromethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(chloromethyl)isoindole-1,3-dione
Molecular Weight (g/mol): 195.6
Molecular Formula: C9H6ClNO2
InChi Key: JKGLRGGCGUQNEX-UHFFFAOYSA-N
SMILES: ClCN1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)