missing translation for 'onlineSavingsMsg'
Learn More

N-(Chloromethyl)phthalimide, 97%

Catalog No. AAA1201814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1201814 25 g
AAA1201822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1201814 Supplier Thermo Scientific Chemicals Supplier No. A1201814

CAS: 17564-64-6 | C9H6ClNO2 | 195.602 g/mol

It is used as an agrochemical and medicine intermediates and is used in organic synthesis. Microporous membranes prepared from chemically modified polysulfone, which is a derivative of this compound have better streaming potential.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an agrochemical and medicine intermediates and is used in organic synthesis. Microporous membranes prepared from chemically modified polysulfone, which is a derivative of this compound have better streaming potential.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.

Chemical Identifiers

CAS 17564-64-6
Molecular Formula C9H6ClNO2
Molecular Weight (g/mol) 195.602
MDL Number MFCD00005898
InChI Key JKGLRGGCGUQNEX-UHFFFAOYSA-N
Synonym n-chloromethyl phthalimide, 2-chloromethyl isoindoline-1,3-dione, n-chloromethylphthalimide, chloromethylphthalimide, n-chloromethyltrimellitimide, 1h-isoindole-1,3 2h-dione, 2-chloromethyl, phthalimide, n-chloromethyl, 2-chloromethyl isoindole-1,3-dione, 2-chloromethyl-1h-isoindole-1,3 2h-dione
PubChem CID 87154
IUPAC Name 2-(chloromethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCl

Specifications

Melting Point 131°C to 135°C
Quantity 25 g
Beilstein 140942
Solubility Information Insoluble in water.
Formula Weight 195.61
Percent Purity 97%
Chemical Name or Material N- (Chloromethyl)phthalimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-541-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only