missing translation for 'onlineSavingsMsg'
Learn More

N-Chloromethyl-4-nitrophthalimide 98.0+%, TCI America™

Catalog No. A55031G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55031G 1 g
A55035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A55031G Supplier TCI America Supplier No. A55031G

Chemical Identifiers

CAS 54455-34-4
Molecular Formula C9H5ClN2O4
Molecular Weight (g/mol) 240.599
MDL Number MFCD00059889
InChI Key YXMUGXUCSVUONH-UHFFFAOYSA-N
Synonym 4-Nitro-N-chloromethylphthalimide
PubChem CID 3016883
IUPAC Name 2-(chloromethyl)-5-nitroisoindole-1,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)N(C2=O)CCl

Specifications

Color Yellow
Quantity 1 g
Formula Weight 240.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chloromethyl-4-nitrophthalimide [for HPLC Labeling]